Learn More
Waters Corp WATERS CORPORATION UPLC HSS C18 Column, 1.8 μm, 100 Å
Catalog No. 50781822 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C18 bonded phase column for ultra-performance liquid chromatography offers superior peak shape, increased retention, and extended column lifetimes at low pH. It is scalable to HPLC separations.
  • High Strength Silica (HSS) particle technology provides increased mechanical stability and retentivity.
  • 1.8 μm HSS particle is specifically designed and tested for UPLC separations.
  • Full end-capped, high coverage C18 ligand provides superior peak shapes for bases and resists acidic hydrolysis for long lifetimes.
  • eCord chip included for tracking column use data, including chemistry type, dimensions, serial number, and performance parameters.
  • Individually tested and includes a performance chromatogram and certificate of batch analysis.
Catalog No. 50-781-822 Supplier Waters Corp Supplier No. 186003532
This C18 bonded phase column for ultra-performance liquid chromatography offers superior peak shape, increased retention, and extended column lifetimes at low pH. It is scalable to HPLC separations.
  • High Strength Silica (HSS) particle technology provides increased mechanical stability and retentivity.
  • 1.8 μm HSS particle is specifically designed and tested for UPLC separations.
  • Full end-capped, high coverage C18 ligand provides superior peak shapes for bases and resists acidic hydrolysis for long lifetimes.
  • eCord chip included for tracking column use data, including chemistry type, dimensions, serial number, and performance parameters.
  • Individually tested and includes a performance chromatogram and certificate of batch analysis.