Learn More
Waters Corp Waters UPLC Column, HSS C18, 100Å Pore, 1.8 μm Particle
Supplier:  Waters Corp 176001819
Encompass_Preferred
This HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase. It provides superior peak shape for bases, increased retention, and extremely long column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations.
  • Provides superior peak shape for bases
  • Increased retention
  • Extremely long column lifetimes at low pH
  • Fully scalable and transferable to HPLC separations
Catalog No. 50-791-052