Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 Column, 100Å Pore, 1.8μm Particle, 3.0 × 150 mm
Catalog No. 50791053 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 Columns provide increased retention for both polar and non-polar analytes. These columns are specifically designed as ACQUITY UPLC HSS Columns and are suitable for operation under UPLC system pressures, offering mechanical strength and stability.
  • Increased retention for both polar and non-polar analytes.
  • Thermally treated silica particle for mechanical strength and stability under UPLC system pressures.
  • Robust particle technology maximizes selectivity space.
  • Enables traditional hydrophobic, reversed-phase interactions.
  • Supports dipole-dipole, aromatic, and hydrogen-bonding interactions.
  • Ideal for general reversed-phase separations.
Catalog No. 50-791-053 Supplier Waters Corp Supplier No. 176001820
ACQUITY UPLC HSS C18 Columns provide increased retention for both polar and non-polar analytes. These columns are specifically designed as ACQUITY UPLC HSS Columns and are suitable for operation under UPLC system pressures, offering mechanical strength and stability.
  • Increased retention for both polar and non-polar analytes.
  • Thermally treated silica particle for mechanical strength and stability under UPLC system pressures.
  • Robust particle technology maximizes selectivity space.
  • Enables traditional hydrophobic, reversed-phase interactions.
  • Supports dipole-dipole, aromatic, and hydrogen-bonding interactions.
  • Ideal for general reversed-phase separations.