Learn More
Waters Corp ACQUITY UPLC HSS C18 Column, 100Å, 1.8 µm, 3 mm X 50 mm, 3/pk
Catalog No. 50791051 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase that provides superior peak shape for bases, increased retention (vs. ACQUITY UPLC BEH C18 columns), and extremely long column lifetimes at low pH. ACQUITY HSS C18 columns are fully scalable and transferable to HPLC separations.
Catalog No. 50-791-051 Supplier Waters Corp Supplier No. 176001818
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase that provides superior peak shape for bases, increased retention (vs. ACQUITY UPLC BEH C18 columns), and extremely long column lifetimes at low pH. ACQUITY HSS C18 columns are fully scalable and transferable to HPLC separations.