Learn More
Waters Corp Waters Column, C18, 100Å, 1.8 μm, 3 mm X 30 mm
Supplier:  Waters Corp 186004682
Encompass_Preferred
This UPLC HSS C18 chemistry provides an ultra-performance, general-purpose C18 bonded phase. It ensures superior peak shape for bases, increased retention, and extended column lifetimes at low pH. These columns are fully scalable and transferable to HPLC separations, utilizing HSS (High Strength Silica) particle technology for high UPLC pressures.
  • Ultra-performance general-purpose C18 bonded phase analytical column.
  • Superior peak shape for bases and increased retention.
  • Extended column life at low pH.
  • Fully scalable and adaptable to HPLC separations.
  • Withstands UPLC pressures using high strength silica particle technology.
  • Improves mechanical stability while maintaining pore volumes equivalent to HPLC silica.
  • Higher retentivity compared to hybrid particle technology.
  • Ideal platform for developing stationary phases with alternative selectivity.
Catalog No. 50-791-190