Learn More
Waters Corp Waters Corporation ACQUITY UPLC HSS C18 Column, 100 Å Pore Size, 1.8 μm Particle Size, 3 mm × 30 mm, 3 Pack
Catalog No. 50791050 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general-purpose C18 bonded phase. It offers superior peak shape for bases, increased retention compared to ACQUITY UPLC BEH C18 columns, and exceptionally long column lifetimes at low pH. These ACQUITY HSS C18 columns are fully scalable and transferable to HPLC separations.
  • Offers superior peak shape for bases
  • Increased retention compared to ACQUITY UPLC BEH C18 columns
  • Exceptionally long column lifetimes at low pH
  • Fully scalable and transferable to HPLC separations
  • 100 Å pore size
  • 1.8 μm particle size
  • 3 mm × 30 mm column dimensions
  • 3 pack
Catalog No. 50-791-050 Supplier Waters Corp Supplier No. 176001817
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general-purpose C18 bonded phase. It offers superior peak shape for bases, increased retention compared to ACQUITY UPLC BEH C18 columns, and exceptionally long column lifetimes at low pH. These ACQUITY HSS C18 columns are fully scalable and transferable to HPLC separations.
  • Offers superior peak shape for bases
  • Increased retention compared to ACQUITY UPLC BEH C18 columns
  • Exceptionally long column lifetimes at low pH
  • Fully scalable and transferable to HPLC separations
  • 100 Å pore size
  • 1.8 μm particle size
  • 3 mm × 30 mm column dimensions
  • 3 pack