Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 Column, 1.8 μm, 3 mm × 75 mm, 100 Å
Catalog No. 50466154 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase that provides superior peak shape for bases, increased retention (vs. ACQUITY UPLC BEH C18 columns), and extremely long column lifetimes at low pH. HSS C18 columns are fully scalable and transferable to HPLC separations.
  • Superior peak shape for bases.
  • Increased retention (vs. BEH C18 columns).
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.
Catalog No. 50-466-154 Supplier Waters Corp Supplier No. 186005618
Add to Cart
Add to Cart
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase that provides superior peak shape for bases, increased retention (vs. ACQUITY UPLC BEH C18 columns), and extremely long column lifetimes at low pH. HSS C18 columns are fully scalable and transferable to HPLC separations.
  • Superior peak shape for bases.
  • Increased retention (vs. BEH C18 columns).
  • Extremely long column lifetimes at low pH.
  • Fully scalable and transferable to HPLC separations.