Learn More
Waters Corp ACQUITY UPLC HSS C18 Column, 100Å, 1.8 µm, 3 mm X 100 mm, 1/pk
Supplier:  Waters Corp 186004684
Encompass_Preferred
ACQUITY UPLC HSS C18 chemistry is an ultra-performance general purpose C18 bonded phase that provides superior peak shape for bases, increased retention (vs. ACQUITY UPLC BEH C18 columns), and extremely long column lifetimes at low pH. ACQUITY HSS C18 columns are fully scalable and transferable to HPLC separations.
Catalog No. 50-791-192