Learn More
Waters Corp UPLC Column, HSS C18 SB, 1.8 μm, 100 Å Pore
Catalog No. 50791055 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This UPLC HSS C18 SB column utilizes High Strength Silica (HSS) particle technology, a mechanically tolerant, silica-based material engineered to withstand high UPLC pressures. It offers increased retentivity and alternate selectivity for various chromatographic applications.
  • High Strength Silica (HSS) particle technology
  • Trifunctional C18 short-bonded phase
  • Designed to withstand UPLC pressures up to 18,000 psi (1241 bar)
  • Offers alternate selectivity compared to hybrid particles
  • Provides increased retentivity
  • 1.8 μm particle size
  • 100 Å pore diameter
  • Silica-based material
Catalog No. 50-791-055 Supplier Waters Corp Supplier No. 176001822
This UPLC HSS C18 SB column utilizes High Strength Silica (HSS) particle technology, a mechanically tolerant, silica-based material engineered to withstand high UPLC pressures. It offers increased retentivity and alternate selectivity for various chromatographic applications.
  • High Strength Silica (HSS) particle technology
  • Trifunctional C18 short-bonded phase
  • Designed to withstand UPLC pressures up to 18,000 psi (1241 bar)
  • Offers alternate selectivity compared to hybrid particles
  • Provides increased retentivity
  • 1.8 μm particle size
  • 100 Å pore diameter
  • Silica-based material