Learn More
Waters Corp ACQUITY UPLC Column, HSS C18 SB, 100Å, 1.8 μm, 3 x 30 mm
Catalog No. 50791054 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations involving complex mixtures of basic and non-basic compounds. These columns offer an alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases. They are fully scalable and transferable to HPLC separations.
  • Used for low pH separations that contain complex mixtures of basic and non-basic compounds.
  • Offers alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases.
  • Fully scalable and transferable to HPLC separations.
Catalog No. 50-791-054 Supplier Waters Corp Supplier No. 176001821
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations involving complex mixtures of basic and non-basic compounds. These columns offer an alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases. They are fully scalable and transferable to HPLC separations.
  • Used for low pH separations that contain complex mixtures of basic and non-basic compounds.
  • Offers alternative analyte selectivity compared to most modern, high coverage, fully endcapped C18 stationary phases.
  • Fully scalable and transferable to HPLC separations.