Learn More
Waters Corp Waters Column, C18, 1.8 μm, 1 mm, 50 mm
Supplier:  Waters Corp 186004114
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations of complex basic and non-basic compounds. They offer alternative analyte selectivity compared to modern, high-coverage, fully endcapped C18 stationary phases. These columns are fully scalable and transferable to HPLC separations.
  • Utilizes HSS particle technology for UPLC pressures and enhanced mechanical stability.
  • Offers higher retentivity and alternative selectivity vs. hybrid technology.
  • General-purpose, silica-based C18 column suitable for various chemical classes.
  • Provides outstanding peak shapes, low-pH stability, and better retention than hybrid C18 columns.
  • Ensures batch-to-batch consistency, low column bleed, ultra-efficiencies, and seamless format conversion.
  • Improves productivity, overcomes lab challenges, and enhances basic chemical peak shape.
Catalog No. 50-791-145