Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 SB Column, 100Å, 1.8 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 176001560
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations of complex basic and non-basic compounds. They provide alternative analyte selectivity compared to most C18 stationary phases and are scalable to HPLC separations. Packing materials are manufactured in a cGMP, ISO 9001 certified plant using ultra-pure reagents and undergo rigorous chromatographic testing to ensure consistent performance.
  • Designed for low pH separations of complex basic and non-basic compounds with alternative analyte selectivity.
  • Fully scalable and transferable to HPLC separations.
  • Features unique non-end-capped C18 chemistry, increasing retention of basic chemicals and reducing non-basic analyte retention.
  • Provides a powerful method development tool for improved selectivity and consistent, reproducible results.
Catalog No. 50-465-902