Learn More
Waters Corp Waters Column, C18, 100Å, 1.8 μm, 3 mm × 150 mm
Catalog No. 50791196 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are utilized for low pH separations involving complex mixtures of basic and non-basic compounds. These columns offer an alternative analyte selectivity compared to most modern, high-coverage, fully endcapped C18 stationary phases, and are fully scalable and transferable to HPLC separations.
  • Designed for predictable and reproducible results.
  • Utilizes non-end-capped, low-coverage silica-based C18 chemistry.
  • Achieves alternative selectivity for compounds affected by silanophilic interactions.
  • Increases retention of basic chemicals due to secondary interactions with leftover silanols.
  • Reduces non-basic analyte retention through ionic repulsion and poor ligand density.
  • Manufactured with ultra-pure reagents in an ISO 9001 and cGMP certified facility.
  • Each batch chromatographically tested with acidic, basic, and neutral analytes.
Catalog No. 50-791-196 Supplier Waters Corp Supplier No. 186004689
ACQUITY UPLC HSS C18 SB columns are utilized for low pH separations involving complex mixtures of basic and non-basic compounds. These columns offer an alternative analyte selectivity compared to most modern, high-coverage, fully endcapped C18 stationary phases, and are fully scalable and transferable to HPLC separations.
  • Designed for predictable and reproducible results.
  • Utilizes non-end-capped, low-coverage silica-based C18 chemistry.
  • Achieves alternative selectivity for compounds affected by silanophilic interactions.
  • Increases retention of basic chemicals due to secondary interactions with leftover silanols.
  • Reduces non-basic analyte retention through ionic repulsion and poor ligand density.
  • Manufactured with ultra-pure reagents in an ISO 9001 and cGMP certified facility.
  • Each batch chromatographically tested with acidic, basic, and neutral analytes.