Learn More
Waters Corp Waters ACQUITY UPLC Column, HSS C18 SB, 1.8 μm, 100 Å, 2.1 mm X 150 mm
Catalog No. 50465904 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are for low pH separations of complex mixtures, offering alternative selectivity to modern, fully endcapped C18 stationary phases. Scalable to HPLC separations.
  • Unique non-end-capped, low-coverage silica-based C18 chemistry for alternate selectivity.
  • Increased silanol activity for greater basic chemical retention.
  • Reduced non-basic analyte retention due to low ligand density and ionic repulsion.
  • Enhances method development flexibility, allowing selectivity changes for bases while maintaining peak shape.
  • Packing materials created specifically for UPLC systems.
  • Manufactured in a cGMP-certified, ISO 9001-certified facility using ultra-pure reagents.
  • Each batch chromatographically tested for excellent, repeatable performance.
  • Operates effectively within a 2 pH to 8 pH range.
Catalog No. 50-465-904 Supplier Waters Corp Supplier No. 176001562
ACQUITY UPLC HSS C18 SB columns are for low pH separations of complex mixtures, offering alternative selectivity to modern, fully endcapped C18 stationary phases. Scalable to HPLC separations.
  • Unique non-end-capped, low-coverage silica-based C18 chemistry for alternate selectivity.
  • Increased silanol activity for greater basic chemical retention.
  • Reduced non-basic analyte retention due to low ligand density and ionic repulsion.
  • Enhances method development flexibility, allowing selectivity changes for bases while maintaining peak shape.
  • Packing materials created specifically for UPLC systems.
  • Manufactured in a cGMP-certified, ISO 9001-certified facility using ultra-pure reagents.
  • Each batch chromatographically tested for excellent, repeatable performance.
  • Operates effectively within a 2 pH to 8 pH range.