Learn More
Waters Corp Waters ACQUITY UPLC HSS C18 SB Column, 1.8 μm, 2.1 mm × 150 mm
Catalog No. 502037667 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations of complex basic and non-basic compound mixtures, offering alternative analyte selectivity and scalability to HPLC separations. They provide faster separations, superior resolution, and greener chromatography, improving sample throughput and sensitivity.
  • Used for low pH separations with complex basic and non-basic compounds.
  • Offers alternative analyte selectivity compared to modern, high coverage, fully endcapped C18 stationary phases.
  • Fully scalable and transferable to HPLC separations.
  • Delivers faster separations, superior resolution, and greener chromatography.
  • Features intermediate C18 ligand density without end-capping, providing shorter retention for non-basic compounds.
  • Functions as a practical method development tool for complex UPLC separations.
Catalog No. 50-203-7667 Supplier Waters Corp Supplier No. 186004120
ACQUITY UPLC HSS C18 SB columns are designed for low pH separations of complex basic and non-basic compound mixtures, offering alternative analyte selectivity and scalability to HPLC separations. They provide faster separations, superior resolution, and greener chromatography, improving sample throughput and sensitivity.
  • Used for low pH separations with complex basic and non-basic compounds.
  • Offers alternative analyte selectivity compared to modern, high coverage, fully endcapped C18 stationary phases.
  • Fully scalable and transferable to HPLC separations.
  • Delivers faster separations, superior resolution, and greener chromatography.
  • Features intermediate C18 ligand density without end-capping, providing shorter retention for non-basic compounds.
  • Functions as a practical method development tool for complex UPLC separations.