Learn More
Waters Corp Waters ACQUITY UPLC Method Validation Kit, HSS C18 SB, 1.8 μm, 100 Å, 2.1 × 50 mm
Supplier:  Waters Corp 186004137
Encompass_Preferred
This ACQUITY UPLC Method Validation Kit - HSS C18 SB provides three columns from different batches for method evaluation, ensuring reproducibility. Its HSS (High Strength Silica) particles enhance retention for diverse analytes, while thermally treated silica ensures stability under UPLC system pressures.
  • HSS (High Strength Silica) particles offer increased retention for polar and non-polar analytes.
  • Thermally treated silica provides mechanical strength and stability under UPLC system pressures.
  • Maximizes selectivity space and supports various interactions: hydrophobic, reversed-phase, dipole-dipole, aromatic, and hydrogen-bonding.
  • HSS C18 SB columns show distinct silanol activity and hydrophobicity within US Pharmacopeia L1 classification.
  • Includes three columns from three batches, ensuring reproducibility and consistency for method evaluation.
Catalog No. 50-466-003