Learn More
Waters Corp Waters Corporation Method Validation Kit, 1.8 μm, 2.1 mm × 150 mm, 3/pk
Catalog No. 50201511 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This kit features three ACQUITY UPLC HSS T3 columns with different sorbent batches, combining analysis columns in one pack. It provides C18 chemistries for diverse applications, enhancing speed, resolution, and sensitivity. BEH Technology improves efficiency, strength, and pH range, enabling rapid method development and robust separations. Designed for ACQUITY UPLC System use.
  • Includes three HSS T3 columns with varied sorbent batches.
  • Combines three analysis columns for comprehensive applications.
  • Features C18 chemistries for broad use.
  • BEH Technology enhances efficiency, strength, and pH range.
  • Designed for ACQUITY UPLC System integration.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Each column individually tested with eCord chip documentation.
Catalog No. 50-201-511 Supplier Waters Corp Supplier No. 186006025
This kit features three ACQUITY UPLC HSS T3 columns with different sorbent batches, combining analysis columns in one pack. It provides C18 chemistries for diverse applications, enhancing speed, resolution, and sensitivity. BEH Technology improves efficiency, strength, and pH range, enabling rapid method development and robust separations. Designed for ACQUITY UPLC System use.
  • Includes three HSS T3 columns with varied sorbent batches.
  • Combines three analysis columns for comprehensive applications.
  • Features C18 chemistries for broad use.
  • BEH Technology enhances efficiency, strength, and pH range.
  • Designed for ACQUITY UPLC System integration.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Each column individually tested with eCord chip documentation.