Learn More
Waters Corp Waters ACQUITY UPLC HSS T3 Column, 1.8 μm, 100 mm
Catalog No. 50780043 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC HSS T3 Column is a universal, silica-based bonded phase column for 100% aqueous mobile phase. Designed for high-speed applications, it features advanced T3 bonding and proprietary end-capping, offering superior performance for separating polar and non-polar compounds across a wide pH range, ensuring predictable column lifetimes.
  • Universal, silica-based bonded phase for 100% aqueous mobile phase, with ultra-low MS bleed.
  • Advanced T3 bonding and end-capping for superior polar and non-polar compound retention and aqueous compatibility.
  • Designed for UPLC applications up to 15,000 psi/1000 bar, tolerating demanding conditions.
  • Offers superior performance and enhanced column performance, peak shape, stability, and predictable lifetimes across a wide pH range.
Catalog No. 50-780-043 Supplier Waters Corp Supplier No. 176001132
The ACQUITY UPLC HSS T3 Column is a universal, silica-based bonded phase column for 100% aqueous mobile phase. Designed for high-speed applications, it features advanced T3 bonding and proprietary end-capping, offering superior performance for separating polar and non-polar compounds across a wide pH range, ensuring predictable column lifetimes.
  • Universal, silica-based bonded phase for 100% aqueous mobile phase, with ultra-low MS bleed.
  • Advanced T3 bonding and end-capping for superior polar and non-polar compound retention and aqueous compatibility.
  • Designed for UPLC applications up to 15,000 psi/1000 bar, tolerating demanding conditions.
  • Offers superior performance and enhanced column performance, peak shape, stability, and predictable lifetimes across a wide pH range.