Learn More
Waters Corp Waters ACQUITY UPLC HSS T3 Column, 100Å, 1.8 μm, 1 mm x 50 mm
Supplier:  Waters Corp 176001127
Encompass_Preferred
ACQUITY HSS T3 sorbents are universal, silica-based bonded phases compatible with 100% aqueous mobile phases, ideal for separating polar and non-polar compounds. These UPLC columns offer ultra-low MS bleed, are scalable to HPLC, and increase speed, sensitivity, resolution, chemical stability, and backpressure resistance.
  • Compatible with 100% aqueous mobile phase
  • Ideal for polar and non-polar compound separations
  • Exhibits ultra-low MS bleed
  • Scalable and transferable to HPLC separations
  • Increases speed, sensitivity, and resolution
  • Maintains chemical stability
  • Features rugged silica particle for extreme backpressures
  • Wide, usable pH range for various analytes
  • Simplifies method transfer
  • Improves chromatographic resolution and granularity
  • Reduces laboratory costs
  • Medium silanol activity
Catalog No. 50-780-039