Learn More
Waters Corp Waters ACQUITY UPLC HSS T3 Column, 100Å, 1.8 μm, 2.1 mm × 100 mm
Catalog No. 50781829 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY UPLC HSS T3 Column is a universal, silica-based bonded phase column designed for developing separations for both polar and non-polar compounds. It offers ultra-low MS bleed and is fully scalable and transferable to HPLC separations. The column provides excellent performance across a wide range of pH and is certified for use in UPLC separations, ensuring robust and reliable analytical results.
  • Universal, silica-based bonded phase, 100% aqueous mobile phase compatible.
  • Separates polar and non-polar compounds.
  • Scalable and transferable to HPLC separations.
  • Excellent performance across a wide pH range (2 to 8 pH).
  • High Strength Silica (HSS) particle for 15,000 psi applications.
  • Advanced T3 bonding for superior retention and aqueous compatibility.
  • C18 chemistry, 100Å pore size, 1.8 μm particle size.
  • Inner diameter 2.1 mm, length 100 mm.
Catalog No. 50-781-829 Supplier Waters Corp Supplier No. 186003539
This ACQUITY UPLC HSS T3 Column is a universal, silica-based bonded phase column designed for developing separations for both polar and non-polar compounds. It offers ultra-low MS bleed and is fully scalable and transferable to HPLC separations. The column provides excellent performance across a wide range of pH and is certified for use in UPLC separations, ensuring robust and reliable analytical results.
  • Universal, silica-based bonded phase, 100% aqueous mobile phase compatible.
  • Separates polar and non-polar compounds.
  • Scalable and transferable to HPLC separations.
  • Excellent performance across a wide pH range (2 to 8 pH).
  • High Strength Silica (HSS) particle for 15,000 psi applications.
  • Advanced T3 bonding for superior retention and aqueous compatibility.
  • C18 chemistry, 100Å pore size, 1.8 μm particle size.
  • Inner diameter 2.1 mm, length 100 mm.