Learn More
Waters Corp Waters Corporation Column, HSS T3, 1.8 μm, 2.1 mm × 30 mm, 100 Å
Catalog No. 50782078 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC HSS T3 Column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, ideal for separating both polar and non-polar compounds. It provides ultra-low MS bleed, is scalable, and transferable to HPLC. The column’s thermally treated High Strength Silica (HSS) particles ensure robustness under high UPLC pressures and enhanced analyte retention.
  • Compatible with 100% aqueous mobile phase.
  • Increased retention for polar and non-polar analytes.
  • Mechanically stable under high UPLC system pressures.
  • Maximizes selectivity space for diverse chromatographic interactions.
  • Seamless integration with UPLC systems.
  • High efficiencies and best-in-class column stability.
  • Facilitates high-throughput method development.
  • Performance verified with Neutrals and Reversed-Phase QC Reference Materials.
Catalog No. 50-782-078 Supplier Waters Corp Supplier No. 186003944
The ACQUITY UPLC HSS T3 Column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, ideal for separating both polar and non-polar compounds. It provides ultra-low MS bleed, is scalable, and transferable to HPLC. The column’s thermally treated High Strength Silica (HSS) particles ensure robustness under high UPLC pressures and enhanced analyte retention.
  • Compatible with 100% aqueous mobile phase.
  • Increased retention for polar and non-polar analytes.
  • Mechanically stable under high UPLC system pressures.
  • Maximizes selectivity space for diverse chromatographic interactions.
  • Seamless integration with UPLC systems.
  • High efficiencies and best-in-class column stability.
  • Facilitates high-throughput method development.
  • Performance verified with Neutrals and Reversed-Phase QC Reference Materials.