Learn More
Waters Corp Waters Column, 100Å Pore Size, 1.8 μm Particle Size, 1 mm ID, 50 mm Length
Catalog No. 50781825 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for separating polar and non-polar compounds. Its advanced T3 bonding and proprietary end-capping provide superior retention and enhanced performance. HSS (High Strength Silica) particle technology ensures mechanical stability under UPLC pressures.
  • Compatible with 100% aqueous mobile phase.
  • Separates polar and non-polar compounds.
  • Ultra-low MS bleed; scalable to HPLC separations.
  • Advanced T3 bonding promotes polar compound retention.
  • Proprietary T3 end-capping enhances performance and stability.
  • HSS particle technology withstands UPLC pressures.
  • Offers increased retentivity.
Catalog No. 50-781-825 Supplier Waters Corp Supplier No. 186003535
This column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phase, suitable for separating polar and non-polar compounds. Its advanced T3 bonding and proprietary end-capping provide superior retention and enhanced performance. HSS (High Strength Silica) particle technology ensures mechanical stability under UPLC pressures.
  • Compatible with 100% aqueous mobile phase.
  • Separates polar and non-polar compounds.
  • Ultra-low MS bleed; scalable to HPLC separations.
  • Advanced T3 bonding promotes polar compound retention.
  • Proprietary T3 end-capping enhances performance and stability.
  • HSS particle technology withstands UPLC pressures.
  • Offers increased retentivity.