Learn More
Waters Corp Waters UPLC Column, C18, 1.8 μm, 100 Å, 1 mm X 100 mm
Catalog No. 50781826 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These UPLC columns feature HSS T3 sorbents with a universal, silica-based bonded phase compatible with 100% aqueous mobile phases. They are ideal for separating both polar and non-polar compounds, offering ultra-low MS bleed and scalability to HPLC separations.
  • C18 chemistry
  • 1.8 μm spherical particle size
  • 100 Å pore size
  • 1 mm × 100 mm dimensions
  • pH range: 2 to 8
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped
  • T3 bonding technology
  • UPLC, UHPLC system compatibility
  • eCord enabled
Catalog No. 50-781-826 Supplier Waters Corp Supplier No. 186003536
These UPLC columns feature HSS T3 sorbents with a universal, silica-based bonded phase compatible with 100% aqueous mobile phases. They are ideal for separating both polar and non-polar compounds, offering ultra-low MS bleed and scalability to HPLC separations.
  • C18 chemistry
  • 1.8 μm spherical particle size
  • 100 Å pore size
  • 1 mm × 100 mm dimensions
  • pH range: 2 to 8
  • Maximum pressure: 18000 psi (1240 Bar)
  • Endcapped
  • T3 bonding technology
  • UPLC, UHPLC system compatibility
  • eCord enabled