Learn More
Waters Corp Waters ACQUITY UPLC HSS T3 Column, 100Å, 1.8 μm, 3 mm x 100 mm
Catalog No. 502037675 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This universal, silica-based bonded phase column is an excellent choice for developing separations of both polar and non-polar compounds, offering full compatibility with 100% aqueous mobile phases.
  • Compatible with 100% aqueous mobile phases.
  • Ideal for separating polar and non-polar compounds.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to HPLC separations.
Catalog No. 50-203-7675 Supplier Waters Corp Supplier No. 186004680
This universal, silica-based bonded phase column is an excellent choice for developing separations of both polar and non-polar compounds, offering full compatibility with 100% aqueous mobile phases.
  • Compatible with 100% aqueous mobile phases.
  • Ideal for separating polar and non-polar compounds.
  • Exhibits ultra-low MS bleed.
  • Fully scalable and transferable to HPLC separations.