Learn More
Waters Corp HSS T3 Column, 100 Å, 1.8 μm, 3 × 100 mm
Catalog No. 50791048 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This universal, silica-based bonded phase is compatible with 100% aqueous mobile phases, ideal for developing separations for both polar and non-polar compounds. These columns exhibit ultra-low MS bleed, scalable and transferable to HPLC separations.
  • Compatible with 100% aqueous mobile phase and ultra-low MS bleed.
  • Scalable and transferable to HPLC separations.
  • Balanced retention for polar and hydrophobic molecules.
  • Trifunctional C18 alkyl phase for enhanced polar compound retention and aqueous compatibility.
  • Advanced T3 bonding and end-capping improves performance, longevity, and stability.
  • High strength silica (HSS) withstands UPLC pressures, improving mechanical stability and retentivity.
  • HSS provides an excellent substrate for alternative selectivity.
Catalog No. 50-791-048 Supplier Waters Corp Supplier No. 176001815
This universal, silica-based bonded phase is compatible with 100% aqueous mobile phases, ideal for developing separations for both polar and non-polar compounds. These columns exhibit ultra-low MS bleed, scalable and transferable to HPLC separations.
  • Compatible with 100% aqueous mobile phase and ultra-low MS bleed.
  • Scalable and transferable to HPLC separations.
  • Balanced retention for polar and hydrophobic molecules.
  • Trifunctional C18 alkyl phase for enhanced polar compound retention and aqueous compatibility.
  • Advanced T3 bonding and end-capping improves performance, longevity, and stability.
  • High strength silica (HSS) withstands UPLC pressures, improving mechanical stability and retentivity.
  • HSS provides an excellent substrate for alternative selectivity.