Learn More
Waters Corp Waters Corporation Column, HSS T3, 1.8 μm, 3 mm, 150 mm
Catalog No. 50791049 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC HSS T3 Column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phases for separating polar and non-polar compounds. It provides ultra-low MS bleed and is scalable to HPLC separations. Utilizing HSS (High Strength Silica) particle technology, it offers increased retentivity and alternate selectivity, simplifying method switching and enhancing productivity.
  • Compatible with 100% aqueous mobile phases for polar and non-polar compound separations.
  • Provides ultra-low MS bleed and is scalable to HPLC separations.
  • Low ligand density enhances retention of polar chemicals and metabolites.
  • Utilizes mechanically tolerant, silica-based HSS particle technology for UPLC pressures.
  • Offers increased retentivity and alternate selectivity, simplifying method switching and enhancing productivity.
Catalog No. 50-791-049 Supplier Waters Corp Supplier No. 176001816
The ACQUITY UPLC HSS T3 Column features a universal, silica-based bonded phase compatible with 100% aqueous mobile phases for separating polar and non-polar compounds. It provides ultra-low MS bleed and is scalable to HPLC separations. Utilizing HSS (High Strength Silica) particle technology, it offers increased retentivity and alternate selectivity, simplifying method switching and enhancing productivity.
  • Compatible with 100% aqueous mobile phases for polar and non-polar compound separations.
  • Provides ultra-low MS bleed and is scalable to HPLC separations.
  • Low ligand density enhances retention of polar chemicals and metabolites.
  • Utilizes mechanically tolerant, silica-based HSS particle technology for UPLC pressures.
  • Offers increased retentivity and alternate selectivity, simplifying method switching and enhancing productivity.