Learn More
Waters Corp ACQUITY UPLC HSS T3 Method Validation Kit, 100Å, 1.8 µm, 3 mm X 50 mm, 3/pk
Supplier:  Waters Corp 186004701
Encompass_Preferred
This kit consists of three ACQUITY UPLC HSS T3 columns packed with three different batches of sorbent.
Catalog No. 50-466-035