Learn More
Waters Corp Tubing Upgrade Kit, CM-A, 2 Column, Stainless Steel
Catalog No. 502392833 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
A tubing kit containing stainless steel tubing assemblies used to implement a CM-A active column manager onto a UPLC or I-Class system for a two-column configuration. This kit can also replace all the stainless steel tubing in this configuration and includes the relevant active preheater.
  • Contains stainless steel tubing assemblies.
  • Designed to implement a CM-A active column manager.
  • Supports a two-column configuration.
  • Can replace all stainless steel tubing in this configuration.
  • Includes the relevant active preheater.
Catalog No. 50-239-2833 Supplier Waters Corp Supplier No. 205000898
A tubing kit containing stainless steel tubing assemblies used to implement a CM-A active column manager onto a UPLC or I-Class system for a two-column configuration. This kit can also replace all the stainless steel tubing in this configuration and includes the relevant active preheater.
  • Contains stainless steel tubing assemblies.
  • Designed to implement a CM-A active column manager.
  • Supports a two-column configuration.
  • Can replace all stainless steel tubing in this configuration.
  • Includes the relevant active preheater.