Learn More
Waters Corp Tubing Kit, UPLC, CM-A, 2 Column, Stainless Steel
Catalog No. 502392209 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This tubing kit contains stainless steel tubing assemblies designed to integrate an active column manager onto a UPLC or I-Class PLUS system for a two-column configuration. It can also be used to replace existing stainless steel tubing in this setup and includes the relevant active preheater.
  • Includes stainless steel tubing assemblies.
  • Designed for two-column configuration.
  • Compatible with ACQUITY UPLC H-Class, I-Class, and M-Class systems.
  • Can be used for performance maintenance or as an upgrade kit.
  • Features an active preheater.
Catalog No. 50-239-2209 Supplier Waters Corp Supplier No. 205001826
This tubing kit contains stainless steel tubing assemblies designed to integrate an active column manager onto a UPLC or I-Class PLUS system for a two-column configuration. It can also be used to replace existing stainless steel tubing in this setup and includes the relevant active preheater.
  • Includes stainless steel tubing assemblies.
  • Designed for two-column configuration.
  • Compatible with ACQUITY UPLC H-Class, I-Class, and M-Class systems.
  • Can be used for performance maintenance or as an upgrade kit.
  • Features an active preheater.