Learn More
Waters Corp Waters M-Class C18 Trap Column, 100Å, 5 μm
Catalog No. 50727106 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC M-Class Symmetry C18 Trap Column is engineered for exceptional performance in nano- to micro-scale liquid chromatographic separations, crucial for biomarker discovery and the identification and characterization of proteins and peptides. These columns are built to withstand high operating pressures of up to 15,000 psi (1034 Bar) and effectively manage pressure fluctuations during injection, thereby ensuring superior longevity and consistent results.
  • Minimize peak broadening while maximizing chromatographic efficiency.
  • Suitable for a variety of 1D (low pH) nanoflow applications.
  • Includes Symmetry 100 Å pore, C18, 5 μm particles.
  • Symmetry silica particles offer the highest retention for peptides, suitable for trapping applications.
  • Connects with two V-detail fittings to the main injection valve.
  • Suitable for bi-directional flow.
Catalog No. 50-727-106 Supplier Waters Corp Supplier No. 186007500
The ACQUITY UPLC M-Class Symmetry C18 Trap Column is engineered for exceptional performance in nano- to micro-scale liquid chromatographic separations, crucial for biomarker discovery and the identification and characterization of proteins and peptides. These columns are built to withstand high operating pressures of up to 15,000 psi (1034 Bar) and effectively manage pressure fluctuations during injection, thereby ensuring superior longevity and consistent results.
  • Minimize peak broadening while maximizing chromatographic efficiency.
  • Suitable for a variety of 1D (low pH) nanoflow applications.
  • Includes Symmetry 100 Å pore, C18, 5 μm particles.
  • Symmetry silica particles offer the highest retention for peptides, suitable for trapping applications.
  • Connects with two V-detail fittings to the main injection valve.
  • Suitable for bi-directional flow.