Learn More
Waters Corp Column, 100Å, 1.8 μm, 100 μm x 100 mm
Catalog No. 502391390 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC M-Class Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility. They feature novel frit technology to withstand 15kpsi operating pressure, ensuring superior longevity.
  • Improves retention of polar chemicals and metabolites in reversed-phase liquid chromatography.
  • Provides balanced retention of polar and hydrophobic compounds without ion-pair reagents.
  • Suitable for examining diverse and complex formulation compositions.
  • HSS (High Strength Silica) particle technology offers improved mechanical stability and higher retentivity.
  • Excellent substrate for producing stationary phases with diverse selectivity.
  • Trifunctional bonding ensures good peak forms, pH stability, and greater retention compared to hybrid-based C18 columns.
Catalog No. 50-239-1390 Supplier Waters Corp Supplier No. 186008008
ACQUITY UPLC M-Class Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility. They feature novel frit technology to withstand 15kpsi operating pressure, ensuring superior longevity.
  • Improves retention of polar chemicals and metabolites in reversed-phase liquid chromatography.
  • Provides balanced retention of polar and hydrophobic compounds without ion-pair reagents.
  • Suitable for examining diverse and complex formulation compositions.
  • HSS (High Strength Silica) particle technology offers improved mechanical stability and higher retentivity.
  • Excellent substrate for producing stationary phases with diverse selectivity.
  • Trifunctional bonding ensures good peak forms, pH stability, and greater retention compared to hybrid-based C18 columns.