Learn More
Waters Corp ACQUITY UPLC M-Class HSS T3 Column, 100Å, 1.8μm, 100 mm
Catalog No. 502391414 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separations, crucial for biomarker discovery and protein/peptide characterization. They operate effectively at 15000 psi (1034 Bar) with excellent resistance to pressure fluctuations. This product utilizes High Strength Silica (HSS) Technology.
  • Optimal for nano- to micro-scale liquid chromatographic separations.
  • Ideal for biomarker discovery and protein/peptide characterization.
  • Operates effectively at 15000 psi (1034 Bar).
  • Excellent resistance to pressure fluctuations, enhancing longevity.
  • High Strength Silica (HSS) Technology with 100 Å pore and 1.8 μm particle size.
  • High retentivity and balanced retention for polar and hydrophobic analytes.
  • T3 bonding for aqueous mobile phase compatibility.
Catalog No. 50-239-1414 Supplier Waters Corp Supplier No. 186008006
These columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separations, crucial for biomarker discovery and protein/peptide characterization. They operate effectively at 15000 psi (1034 Bar) with excellent resistance to pressure fluctuations. This product utilizes High Strength Silica (HSS) Technology.
  • Optimal for nano- to micro-scale liquid chromatographic separations.
  • Ideal for biomarker discovery and protein/peptide characterization.
  • Operates effectively at 15000 psi (1034 Bar).
  • Excellent resistance to pressure fluctuations, enhancing longevity.
  • High Strength Silica (HSS) Technology with 100 Å pore and 1.8 μm particle size.
  • High retentivity and balanced retention for polar and hydrophobic analytes.
  • T3 bonding for aqueous mobile phase compatibility.