Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp UPLC Column, Peptide BEH C18, 300Å, 1.7 μm

Catalog No. 502392960 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These UPLC M-Class columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation, featuring patented BEH Technology 300 Å pore, C18, 1.7μm particles for high retentivity and peak shape.

  • Optimal for nano- to micro-scale liquid chromatographic separation.
  • Ideal for biomarker discovery and protein/peptide identification.
  • Withstands 15000 psi (1034 bar) operating pressure for longevity.
  • Features BEH technology: 300 Å pore, C18, 1.7 μm particles.
  • Offers highest retentivity for basic compounds and excellent peak shape.
  • Suitable for separations up to 80°C and pH range of 1 to 10.
  • QC tested for peptide separation.
  • Dimensions: 100 μm inner diameter; 100 mm length.

Catalog No. 50-239-2960 Supplier Waters Corp Supplier No. 186007488
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These UPLC M-Class columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation, featuring patented BEH Technology 300 Å pore, C18, 1.7μm particles for high retentivity and peak shape.

  • Optimal for nano- to micro-scale liquid chromatographic separation.
  • Ideal for biomarker discovery and protein/peptide identification.
  • Withstands 15000 psi (1034 bar) operating pressure for longevity.
  • Features BEH technology: 300 Å pore, C18, 1.7 μm particles.
  • Offers highest retentivity for basic compounds and excellent peak shape.
  • Suitable for separations up to 80°C and pH range of 1 to 10.
  • QC tested for peptide separation.
  • Dimensions: 100 μm inner diameter; 100 mm length.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.