Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, C18, 1.7 μm, 75 μm X 150 mm

Catalog No. 502392959 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

ACQUITY UPLC M-Class Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separations, critical for biomarker discovery, and protein and peptide identification and characterization. These columns withstand 15 kpsi (1,000 bar) operating pressure, demonstrating superior longevity and managing pressure fluctuations during injection effectively. This offering incorporates Waters' patented CSH Technology with 130 Å pore, C18, 1.7μm particles, providing enhanced selectivity and exceptional peak shape, especially for basic compounds under low pH conditions.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 10
  • Maximum pressure: 15000 psi (1034 bar)
  • Particle size: 1.7 μm
  • Pore size: 300 Å
  • Inner diameter: 75 μm
  • Length: 150 mm
  • Application: Peptide

Catalog No. 50-239-2959 Supplier Waters Corp Supplier No. 186007490
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACQUITY UPLC M-Class Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separations, critical for biomarker discovery, and protein and peptide identification and characterization. These columns withstand 15 kpsi (1,000 bar) operating pressure, demonstrating superior longevity and managing pressure fluctuations during injection effectively. This offering incorporates Waters' patented CSH Technology with 130 Å pore, C18, 1.7μm particles, providing enhanced selectivity and exceptional peak shape, especially for basic compounds under low pH conditions.

  • Chemistry: C18
  • Separation mode: Reversed Phase
  • Particle substrate: Hybrid
  • pH range: 1 to 10
  • Maximum pressure: 15000 psi (1034 bar)
  • Particle size: 1.7 μm
  • Pore size: 300 Å
  • Inner diameter: 75 μm
  • Length: 150 mm
  • Application: Peptide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.