Learn More
Waters Corp Column, ACQUITY UPLC M-Class Peptide BEH C18, 130Å, 1.7 μm, 75 μm x 200 mm
Catalog No. 502390657 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is engineered for optimal performance in nano- to micro-scale liquid chromatographic separations, offering superior sensitivity, resolution, and reproducibility. It is crucial for biomarker discovery, protein identification, and characterization. Featuring novel frit technology, it withstands high operating pressures up to 15,000 psi (1034 Bar) and manages pressure fluctuations, ensuring exceptional longevity.
  • Optimal for nano- to micro-scale liquid chromatographic separation.
  • High sensitivity, resolution, and reproducibility for biomarker/protein characterization.
  • Novel frit technology; withstands 15,000 psi (1034 Bar) operating pressure, ensuring longevity.
  • Patented BEH Technology: 130Å pore, C18, 1.7μm particles.
  • QC tested for peptide separation using tryptic digest of cytochrome c.
  • Manufactured in ISO 9001 certified plant.
Catalog No. 50-239-0657 Supplier Waters Corp Supplier No. 186007483
This column is engineered for optimal performance in nano- to micro-scale liquid chromatographic separations, offering superior sensitivity, resolution, and reproducibility. It is crucial for biomarker discovery, protein identification, and characterization. Featuring novel frit technology, it withstands high operating pressures up to 15,000 psi (1034 Bar) and manages pressure fluctuations, ensuring exceptional longevity.
  • Optimal for nano- to micro-scale liquid chromatographic separation.
  • High sensitivity, resolution, and reproducibility for biomarker/protein characterization.
  • Novel frit technology; withstands 15,000 psi (1034 Bar) operating pressure, ensuring longevity.
  • Patented BEH Technology: 130Å pore, C18, 1.7μm particles.
  • QC tested for peptide separation using tryptic digest of cytochrome c.
  • Manufactured in ISO 9001 certified plant.