Learn More
Waters Corp Waters Column, Peptide BEH C18, 300Å, 1.7 μm, 150 μm x 100 mm
Catalog No. 502038064 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
ACQUITY UPLC M-Class Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separations, aiding in biomarker discovery, protein, and peptide identification. These columns feature patented BEH Technology with 300 Å pore, C18, 1.7 μm particles, offering high retentivity for basic compounds and excellent peak shape at elevated pH mobile phases. They are suitable for separations up to 80°C and operate at 15 kpsi (1,000 bar).
  • Each batch is QC tested using a gradient separation of a tryptic digest of cytochrome c for peptide separation suitability.
  • Produced with a unique frit method for high-pressure endurance and pressure change management.
  • Individually inspected to meet strict quality standards.
  • Manufactured in an ISO 9001 certified facility.
Catalog No. 50-203-8064 Supplier Waters Corp Supplier No. 186007489
Add to Cart
Add to Cart
ACQUITY UPLC M-Class Columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separations, aiding in biomarker discovery, protein, and peptide identification. These columns feature patented BEH Technology with 300 Å pore, C18, 1.7 μm particles, offering high retentivity for basic compounds and excellent peak shape at elevated pH mobile phases. They are suitable for separations up to 80°C and operate at 15 kpsi (1,000 bar).
  • Each batch is QC tested using a gradient separation of a tryptic digest of cytochrome c for peptide separation suitability.
  • Produced with a unique frit method for high-pressure endurance and pressure change management.
  • Individually inspected to meet strict quality standards.
  • Manufactured in an ISO 9001 certified facility.