Learn More
Waters Corp ACQUITY UPLC M-Class Trap Column, C18, 5 μm, 100 Å, 180 μm x 20 mm
Catalog No. 502390584 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This ACQUITY UPLC M-Class trap column is engineered for optimal performance in nano- to micro-scale liquid chromatographic separations. It provides high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization. Designed for high throughput applications, it facilitates complex 2D separations by enriching and refocusing samples before elution to the nano-separation column.
  • Allows for operation at 15kpsi with robust pressure handling, ensuring longevity.
  • Supports 2D applications using high pH in the first dimension with unique fritting technologies.
  • Endures pressure changes during injection, enhancing longevity.
  • Minimizes peak tailing and maintains efficiency with proper connections.
  • Features a V-detail connection on both ends.
  • Contains 100Å pore, C18, 5μm particles.
Catalog No. 50-239-0584 Supplier Waters Corp Supplier No. 186007592
This ACQUITY UPLC M-Class trap column is engineered for optimal performance in nano- to micro-scale liquid chromatographic separations. It provides high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization. Designed for high throughput applications, it facilitates complex 2D separations by enriching and refocusing samples before elution to the nano-separation column.
  • Allows for operation at 15kpsi with robust pressure handling, ensuring longevity.
  • Supports 2D applications using high pH in the first dimension with unique fritting technologies.
  • Endures pressure changes during injection, enhancing longevity.
  • Minimizes peak tailing and maintains efficiency with proper connections.
  • Features a V-detail connection on both ends.
  • Contains 100Å pore, C18, 5μm particles.