Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Trap Column, C18, 5 μm, 180 μm x 20 mm, 100 Å

Catalog No. 502038070 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These trap columns are for nano- to micro-scale liquid chromatographic separation, ideal for biomarker discovery, protein and peptide identification. They withstand 15 kpsi (1,000 bar) operating pressure, ensuring longevity. They enable complex 2D separations, enriching samples before final separation.

  • High pH compatibility.
  • 100 Å pore, C18, 5 μm spherical silica for high peptide retention.
  • Bi-directional flow, reversed phase separation (pH 2 to 8).
  • Endcapped, low silanol activity.
  • Endfitting: Waters(male)/Valco(male).
  • Dimensions: 180 μm ID, 20 mm length.
  • cGMP, ISO 9001-certified packing.
  • Chromatographically tested with performance chromatogram.
  • Manufactured to exacting specifications with special frit technique.

Catalog No. 50-203-8070 Supplier Waters Corp Supplier No. 186007497
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These trap columns are for nano- to micro-scale liquid chromatographic separation, ideal for biomarker discovery, protein and peptide identification. They withstand 15 kpsi (1,000 bar) operating pressure, ensuring longevity. They enable complex 2D separations, enriching samples before final separation.

  • High pH compatibility.
  • 100 Å pore, C18, 5 μm spherical silica for high peptide retention.
  • Bi-directional flow, reversed phase separation (pH 2 to 8).
  • Endcapped, low silanol activity.
  • Endfitting: Waters(male)/Valco(male).
  • Dimensions: 180 μm ID, 20 mm length.
  • cGMP, ISO 9001-certified packing.
  • Chromatographically tested with performance chromatogram.
  • Manufactured to exacting specifications with special frit technique.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.