Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation MS Start-up Solution Kit, ACQUITY/Quattro micro/Premier, 24 Month Shelf Life

Catalog No. 50792797 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This Mass Spectrometry standards kit is intended for system assessment. It comprises three containers, two ampules and one Nalgene bottle, each holding 10 mL of standard solutions.

  • Designed for ACQUITY, Quattro micro, or Quattro Premier Mass Spectrometry systems.
  • Includes a Certificate of Analysis.
  • 24 month shelf life and room temperature storage.
  • Solution 1 contains 1.0000 mg/mL Sulfadimethoxine in MS Grade Methanol.
  • Solution 2 contains Sulfadimethoxine, Terfenadine, Reserpine, Acetaminophen, and Caffeine (1.0000 mg/mL each) in MS Grade Acetonitrile.
  • Solution 3 consists of Formic Acid, ACS reagent 96% in a 15 mL Brown Nalgene Bottle.
  • Separation mode: Reversed Phase; Detection method: MS.
  • Application for system performance and calibration.
  • Contains hazardous material, requiring special freight handling and possible additional charges.

Catalog No. 50-792-797 Supplier Waters Corp Supplier No. 700002741
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This Mass Spectrometry standards kit is intended for system assessment. It comprises three containers, two ampules and one Nalgene bottle, each holding 10 mL of standard solutions.

  • Designed for ACQUITY, Quattro micro, or Quattro Premier Mass Spectrometry systems.
  • Includes a Certificate of Analysis.
  • 24 month shelf life and room temperature storage.
  • Solution 1 contains 1.0000 mg/mL Sulfadimethoxine in MS Grade Methanol.
  • Solution 2 contains Sulfadimethoxine, Terfenadine, Reserpine, Acetaminophen, and Caffeine (1.0000 mg/mL each) in MS Grade Acetonitrile.
  • Solution 3 consists of Formic Acid, ACS reagent 96% in a 15 mL Brown Nalgene Bottle.
  • Separation mode: Reversed Phase; Detection method: MS.
  • Application for system performance and calibration.
  • Contains hazardous material, requiring special freight handling and possible additional charges.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.