Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters ACQUITY UPLC Column, C18, 1.7 μm, 130 Å

Catalog No. 50782081 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These columns are designed for the separation of detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography. They are QC tested to ensure repeatable performance for demanding oligonucleotide assays. These columns are ideally suited for characterizing oligonucleotides, delivering outstanding peak shape, sample resolution, and extended column life.

  • Optimized for ion-pair, reversed-phase chromatography.
  • Particle size: 1.7 μm.
  • Pore size: 130 Å.
  • Wide pH range: 1 to 12.
  • Hybrid particle substrate with BEH technology for chemical stability.
  • Ideal for UPLC and UHPLC systems.
  • Manufactured under cGMP, ISO 9001 conditions.
  • Includes Certificate of Analysis and Performance Test Chromatogram.
  • Dimensions: 2.1 mm inner diameter x 50 mm length.

Catalog No. 50-782-081 Supplier Waters Corp Supplier No. 186003949
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These columns are designed for the separation of detritylated synthetic oligonucleotides using ion-pair, reversed-phase chromatography. They are QC tested to ensure repeatable performance for demanding oligonucleotide assays. These columns are ideally suited for characterizing oligonucleotides, delivering outstanding peak shape, sample resolution, and extended column life.

  • Optimized for ion-pair, reversed-phase chromatography.
  • Particle size: 1.7 μm.
  • Pore size: 130 Å.
  • Wide pH range: 1 to 12.
  • Hybrid particle substrate with BEH technology for chemical stability.
  • Ideal for UPLC and UHPLC systems.
  • Manufactured under cGMP, ISO 9001 conditions.
  • Includes Certificate of Analysis and Performance Test Chromatogram.
  • Dimensions: 2.1 mm inner diameter x 50 mm length.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.