Learn More
Waters Corp ACQUITY UPLC Column, C18, 1.7 μm, 130 Å, 2.1 mm X 150 mm
Supplier:  Waters Corp 186005516
Encompass_Preferred
This column is designed for exceptional peak shape, sample resolution, and extended column life, suitable for oligonucleotide characterization via ion-pair, reversed-phase chromatography. It resolves large oligonucleotide sequences using sub-3 μm BEH Technology particles within UPLC systems.
  • Each column undergoes QC testing for consistent performance in demanding oligonucleotide assays.
  • Simplifies UPLC analysis of synthetic oligonucleotides.
  • Efficient ion-pairing reversed-phase (IR-RP) chromatography for detritylated synthetic oligonucleotide species.
  • High level of synthetic oligonucleotide purification without compromising recovery.
  • Suitable for purifying and analyzing sensitive DNA or RNA-based oligonucleotide products.
  • Superior packed bed stability under increased temperatures and high pH conditions.
  • Operating pH range of 1–12.
Catalog No. 50-466-131