Learn More
Waters Corp ACQUITY UPLC Peptide Column Kit, 1.7 μm, 1 mm x 150 mm
Catalog No. 502392031 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters ACQUITY UPLC CSH130 C18 1.7 μm particles offer excellent peak shape for peptides in LC or LC-MS applications, even with FA- or TFA-containing mobile phases. This column kit also includes a Cytochrome C Digestion Standard for immediate benchmarking and continuous monitoring, with each batch quality-controlled.
  • Particles contain a low and precisely defined concentration of positive charges.
  • Performance shows little dependence on strong ion pairing agents.
  • Includes one CSH130 C18 column and one vial of Cytochrome C Digestion Standard.
Catalog No. 50-239-2031 Supplier Waters Corp Supplier No. 176003063
Waters ACQUITY UPLC CSH130 C18 1.7 μm particles offer excellent peak shape for peptides in LC or LC-MS applications, even with FA- or TFA-containing mobile phases. This column kit also includes a Cytochrome C Digestion Standard for immediate benchmarking and continuous monitoring, with each batch quality-controlled.
  • Particles contain a low and precisely defined concentration of positive charges.
  • Performance shows little dependence on strong ion pairing agents.
  • Includes one CSH130 C18 column and one vial of Cytochrome C Digestion Standard.