Learn More
Waters Corp ACQUITY UPLC Column, C18, 1.7 μm, 2.1 mm x 50 mm, 3 pk
Supplier: Waters Corp 176003067
The column's 1.7 μm particle column provides excellent peak shape for peptides using FA or TFA mobile phases. Its performance is largely independent of strong ion pairing agents, making it suitable for LC or LC-MS applications. The kit includes one column and a Cytochrome C Digestion Standard for immediate benchmarking and continuous monitoring. Each standard batch is quality controlled with a certificate of analysis.
- Chemistry: C18
- Separation mode: Reversed phase
- pH range: 1–11
- Endcapped: Yes
- Molecular weight range: 1000–15000
- Particle shape: Spherical
- Particle size: 1.7 μm
- Pore size: 130 Å
- QC tested: Peptide
- Particle technology: CSH
- Inner diameter: 2.1 mm
- Length: 50 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.