Learn More
Waters Corp ACQUITY UPLC Peptide CSH C18 Column, 130 Å, 1.7 μm, 2.1 × 50 mm
Catalog No. 502390539 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These ACQUITY UPLC Peptide CSH C18 columns are designed to provide superior peak shape for basic analytes when used with formic acid or other low-ionic-strength, acidic mobile phases, eliminating the need for ion-pairing reagents. Their controlled, low-level, positive surface charge, bonded to ethylene-bridged hybrid (BEH) particles, enables this characteristic performance, offering an exception to typical reversed-phase limitations.
  • Superior peak shape for basic analytes.
  • Optimized for formic acid and low-ionic-strength acidic mobile phases.
  • Eliminates the need for ion-pairing reagents.
  • Features a controlled, low-level positive surface charge.
  • Utilizes ethylene-bridged hybrid (BEH) particles.
  • Ideal for method validation.
  • Includes a Cytochrome c Digestion Standard vial.
Catalog No. 50-239-0539 Supplier Waters Corp Supplier No. 176003064
These ACQUITY UPLC Peptide CSH C18 columns are designed to provide superior peak shape for basic analytes when used with formic acid or other low-ionic-strength, acidic mobile phases, eliminating the need for ion-pairing reagents. Their controlled, low-level, positive surface charge, bonded to ethylene-bridged hybrid (BEH) particles, enables this characteristic performance, offering an exception to typical reversed-phase limitations.
  • Superior peak shape for basic analytes.
  • Optimized for formic acid and low-ionic-strength acidic mobile phases.
  • Eliminates the need for ion-pairing reagents.
  • Features a controlled, low-level positive surface charge.
  • Utilizes ethylene-bridged hybrid (BEH) particles.
  • Ideal for method validation.
  • Includes a Cytochrome c Digestion Standard vial.