Learn More
Waters Corp ACQUITY UPLC Peptide CSH C18 Column Kit, 1.0 x 100 mm, 1.7 to 1.8 μm Particle Size, 130Å Pore Size
Catalog No. 502390609 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC Peptide CSH C18 column kit is designed for high-performance liquid chromatography applications. This kit features a column with specific dimensions, particle size, and pore size tailored for peptide analysis.
  • Column dimensions of 1.0 x 100 mm
  • Particle size of 1.7 to 1.8 μm
  • Pore size of 130Å
  • Kit includes a Peptide CSH C18, 130Å column plus one vial of Cytochrome C Digestion Standard.
Catalog No. 50-239-0609 Supplier Waters Corp Supplier No. 176003062
Add to Cart
Add to Cart
The ACQUITY UPLC Peptide CSH C18 column kit is designed for high-performance liquid chromatography applications. This kit features a column with specific dimensions, particle size, and pore size tailored for peptide analysis.
  • Column dimensions of 1.0 x 100 mm
  • Particle size of 1.7 to 1.8 μm
  • Pore size of 130Å
  • Kit includes a Peptide CSH C18, 130Å column plus one vial of Cytochrome C Digestion Standard.