Learn More
Waters Corp ACQUITY UPLC Peptide CSH C18 Column Kit, 130Å, 1.7 μm, 2.1 mm × 150 mm
Catalog No. 502391250 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters ACQUITY UPLC CSH130 C18 1.7 μm particles contain a low and carefully-defined concentration of positive charges. These characteristics yield excellent peak shape for peptides when using either FA- or TFA-containing mobile phases. The column's performance shows little dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • This kit includes one CSH130 C18 column and one vial of Cytochrome C Digestion Standard.
  • The standard is provided for immediate benchmarking upon receipt and can be reordered separately for continuous monitoring.
  • Each batch of the digestion standard is prepared under strict quality control criteria, confirmed by an included certificate of analysis.
Catalog No. 50-239-1250 Supplier Waters Corp Supplier No. 176003066
Waters ACQUITY UPLC CSH130 C18 1.7 μm particles contain a low and carefully-defined concentration of positive charges. These characteristics yield excellent peak shape for peptides when using either FA- or TFA-containing mobile phases. The column's performance shows little dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • This kit includes one CSH130 C18 column and one vial of Cytochrome C Digestion Standard.
  • The standard is provided for immediate benchmarking upon receipt and can be reordered separately for continuous monitoring.
  • Each batch of the digestion standard is prepared under strict quality control criteria, confirmed by an included certificate of analysis.