Learn More
Waters Corp Waters Corporation Column, C18, 1.7 μm, 130 Å, 1 mm x 150 mm
Supplier:  Waters Corp 186006935
Encompass_Preferred
This CSH C18 column utilizes 1.7 μm particles for excellent peptide peak shape, suitable for LC or LC-MS applications with minimal ion pairing agent dependence. Built on the Charged Surface Hybrid (CSH) platform, it enhances loading capacity and provides superior peak shape for basic chemicals under low pH, low ionic strength conditions. CSH technology allows TFA-like peak shapes using MS-friendly mobile phases, streamlining method development.
  • Excellent peak shape for peptides.
  • Ideal for LC or LC-MS applications.
  • Offers alternate selectivity.
  • Enhanced loading capacity for basic chemicals.
  • Reduced column bleed and high efficiency.
  • Enables robust and transferable procedures.
  • Simulates TFA peak shape without using TFA.
  • Facilitates faster and easier technique development.
Catalog No. 50-203-7770