Learn More
Waters Corp ACQUITY UPLC Column, SEC, 2.5 μm, 200 Å, 4.6 mm × 50 mm
Supplier:  Waters Corp 186009082
Encompass_Preferred
This ACQUITY UPLC Column is designed for legacy methods and is the first commercially available sub-2-μm SEC chemistry. It is optimized for the analysis of proteins and their aggregates with molecular weights ranging from 10,000 to 450,000 Daltons, providing fast and accurate aggregate determinations up to 10 times quicker than traditional HPLC-based SEC methods.
  • First commercially available sub-2-μm SEC chemistry.
  • Optimized for the analysis of proteins and their aggregates with molecular weights ranging from 10,000 to 450,000 Daltons.
  • Delivers fast, accurate aggregate determinations up to 10 times quicker than traditional HPLC-based SEC methods.
  • Primarily supports legacy methods.
Catalog No. 50-152-8616