Learn More
Waters Corp Waters ACQUITY UPLC Protein BEH SEC Column, 200 Å, 1.7 μm, 4.6 mm x 150 mm
Catalog No. 501461891 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is the first commercially available sub-2-μm SEC chemistry, optimized for analyzing proteins and their aggregates with molecular weights from 10,000 to 450,000 Daltons. It delivers fast, accurate aggregate determinations, up to 10 times quicker than traditional HPLC methods, for optimal UPLC SEC separation.
  • Features Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface for stable chemistry.
  • Manufactured in a cGMP, ISO 9001 certified plant with tested batches for reproducible performance.
  • eCord intelligent chip technology tracks lifetime performance and captures data automatically.
  • Recommended pH range: 2 to 8; operating temperature: 4°C to 60°C.
  • Maximum recommended pressure: 7000 PSI (483 Bar).
  • Particle size: 1.7 μm; pore size: 200 Å; dimensions: 4.6 mm x 150 mm.
Catalog No. 50-146-1891 Supplier Waters Corp Supplier No. 176003904
This column is the first commercially available sub-2-μm SEC chemistry, optimized for analyzing proteins and their aggregates with molecular weights from 10,000 to 450,000 Daltons. It delivers fast, accurate aggregate determinations, up to 10 times quicker than traditional HPLC methods, for optimal UPLC SEC separation.
  • Features Ethylene Bridged Hybrid (BEH) particle technology and a diol-bonded surface for stable chemistry.
  • Manufactured in a cGMP, ISO 9001 certified plant with tested batches for reproducible performance.
  • eCord intelligent chip technology tracks lifetime performance and captures data automatically.
  • Recommended pH range: 2 to 8; operating temperature: 4°C to 60°C.
  • Maximum recommended pressure: 7000 PSI (483 Bar).
  • Particle size: 1.7 μm; pore size: 200 Å; dimensions: 4.6 mm x 150 mm.