Learn More
Waters Corp Waters ACQUITY UPLC Protein BEH SEC Column, 125Å, 1.7 μm, 4.6 mm x 300 mm
Catalog No. 501461894 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The ACQUITY UPLC Protein BEH SEC Column is the first sub-2-μm SEC chemistry, optimized for analyzing proteins and their aggregates with molecular weights from 1,000 to 80,000 Daltons. It provides fast, accurate aggregate determinations up to 10 times quicker than traditional HPLC-based SEC methods.
  • Optimized for protein and aggregate analysis.
  • Molecular weight range: 1,000 to 80,000 daltons.
  • Delivers fast, accurate aggregate determinations.
  • Improves resolution, throughput, and sensitivity.
  • Designed for UPLC SEC applications.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Includes a protein standard mix for benchmarking.
Catalog No. 50-146-1894 Supplier Waters Corp Supplier No. 176003907
Add to Cart
Add to Cart
Request a Quote
The ACQUITY UPLC Protein BEH SEC Column is the first sub-2-μm SEC chemistry, optimized for analyzing proteins and their aggregates with molecular weights from 1,000 to 80,000 Daltons. It provides fast, accurate aggregate determinations up to 10 times quicker than traditional HPLC-based SEC methods.
  • Optimized for protein and aggregate analysis.
  • Molecular weight range: 1,000 to 80,000 daltons.
  • Delivers fast, accurate aggregate determinations.
  • Improves resolution, throughput, and sensitivity.
  • Designed for UPLC SEC applications.
  • Manufactured in a cGMP, ISO 9001 certified plant.
  • Includes a protein standard mix for benchmarking.